missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals O-Acetyl-L-serine hydrochloride

Product Code. 15496649
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496649 5 g 5g
15486649 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496649 Supplier Thermo Scientific Chemicals Supplier No. J64933.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Substrate for cysteine synthase and beta-substituted alanine synthase in higher plants | CAS: 66638-22-0 | C5H10ClNO4 | 183.588 g/mol

O-Acetyl-L-serine, a cysteine precursor, is used as a substrate for the identification, differentiation and characterization of O-acetyl-L-serine(thiol)lyase(s) (OASTL) involved in cysteine biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Acetyl-L-serine, a cysteine precursor, is used as a substrate for the identification, differentiation and characterization of O-acetyl-L-serine(thiol)lyase(s) (OASTL) involved in cysteine biosynthesis.

Solubility
Soluble in ethanol almost transparency.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66638-22-0
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00060169
InChI Key MGQOSZSPKMBSRW-WCCKRBBISA-N
Synonym o-acetyl-l-serine hydrochloride, s-3-acetoxy-2-aminopropanoic acid hydrochloride, o-acetyl-l-serine hcl, h-ser ac-oh.hcl, o-acetyl-l-serine-hydrogen chloride 1/1
PubChem CID 44630053
IUPAC Name (2S)-3-acetyloxy-2-aminopropanoic acid;hydrochloride
SMILES CC(=O)OCC(C(=O)O)N.Cl

Specifications

Color White
Solubility Information Soluble in ethanol almost transparency.
Quantity 5 g
Formula Weight 183.59
Physical Form Powder
Chemical Name or Material O-Acetyl-L-serine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.