missing translation for 'onlineSavingsMsg'
Learn More

o-Anisoyl chloride, 97%

Product Code. 10124780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10124780 5 g Glass bottle 5g
10349440 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10124780 Supplier Thermo Scientific Acros Supplier No. 276110050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21615-34-9 | C8H7ClO2 | 170.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21615-34-9
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.6
MDL Number MFCD00000664
InChI Key RZNHSEZOLFEFGB-UHFFFAOYSA-N
Synonym o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi
PubChem CID 88969
IUPAC Name 2-methoxybenzoyl chloride
SMILES COC1=CC=CC=C1C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.1460g/mL
Boiling Point 128°C to 129°C (8.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4COCl
Refractive Index 1.5714 to 1.5734
Quantity 5 g
Beilstein 10, 140
Specific Gravity 1.146
Solubility Information Solubility in water: may decompose in water
Formula Weight 170.6
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material o-Anisoyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • o-Anisoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.