missing translation for 'onlineSavingsMsg'
Learn More

o-Benzoic sulfimide sodium salt hydrate, 99%

Product Code. 11442957
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442957 5000 g 5000g
11432957 250 g 250g
11422957 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442957 Supplier Thermo Scientific Alfa Aesar Supplier No. A15530.0I
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82385-42-0 | C7H4NNaO3S | 205.16 g/mol

O-benzoic sulfimide sodium salt is used as an artificial sweetener in drinks, candies, cookies, medicines and toothpaste. It is used in the food industry and for sweetening diabetes diet. It is also used as a synthetic sweet ignorance agent. It finds application in a variety of beverages, foods, cosmetics and pharmaceuticals to provide sweetness without adding calories. It is involved as a nickel electroplating brightener, chemical intermediate, animal feed sweetener and anaerobic adhesive accelerator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-benzoic sulfimide sodium salt is used as an artificial sweetener in drinks, candies, cookies, medicines and toothpaste. It is used in the food industry and for sweetening diabetes diet. It is also used as a synthetic sweet ignorance agent. It finds application in a variety of beverages, foods, cosmetics and pharmaceuticals to provide sweetness without adding calories. It is involved as a nickel electroplating brightener, chemical intermediate, animal feed sweetener and anaerobic adhesive accelerator.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82385-42-0
Molecular Formula C7H4NNaO3S
Molecular Weight (g/mol) 205.16
MDL Number MFCD00149605
InChI Key OAZGZMKGRPRHJX-UHFFFAOYSA-M
Synonym saccharin sodium salt hydrate
PubChem CID 131673955
IUPAC Name 1,1-dioxo-1,2-benzothiazol-3-one;molecular hydrogen;sodium;hydrate
SMILES [Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12

Specifications

Melting Point >300°C
Quantity 5000 g
Beilstein 3599229
Merck Index 14,8312
Solubility Information Soluble in water.
Formula Weight 205.16 (Anhydrous)
Percent Purity 99%
Chemical Name or Material o-Benzoic sulfimide sodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.