missing translation for 'onlineSavingsMsg'
Learn More

O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate, 98%, Thermo Scientific™

Product Code. 11976691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11976691 25 g 25g
11966691 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976691 Supplier Thermo Scientific Acros Supplier No. 440250250
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 105379-24-6
Molecular Formula C15H20N5O·F6P
Molecular Weight (g/mol) 431.32
InChI Key XKTRAGMCMJYRRN-UHFFFAOYSA-N
Synonym hbpyu, benzotriazol-1-yloxy dipyrrolidinocarbenium hexafluorophosphate, 1h-benzo d 1,2,3 triazol-1-yl di pyrrolidin-1-yl methylene oxonium hexafluorophosphate v, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, hbpyu, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy benzotriazole hexafluorophosphate, 1-1,2,3-benzotriazol-1-yloxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, benzotriazol-1-yloxy dipyrrolidinocarbenium hexaf, 1,2,3-benzotriazol-1-yloxy bis pyrrolidin-1-yl methylium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy benzotriazole hexafluorophosphate
PubChem CID 13685355
IUPAC Name 1-[pyrrolidin-1-ium-1-ylidene(pyrrolidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
SMILES C1CCN(C1)C(=[N+]2CCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 176°C
Color White
Elemental Analysis 40.73 to 42.81% C, 15.83 to 16.64% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 25 g
Formula Weight 431.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.