missing translation for 'onlineSavingsMsg'
Learn More

O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%

Product Code. 15294694 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15294694 5 g Glass bottle 5g
15284694 1 g Glass bottle 1g
15204704 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15294694 Supplier Thermo Scientific Acros Supplier No. 299430050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 94790-37-1 | C11H16F6N5OP | 379.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94790-37-1
InChI Key UQYZFNUUOSSNKT-UHFFFAOYSA-N
Synonym hbtu, o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, 2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, hbtu-reagent, o-benzotriazol-1-yl-tetramethyluronium hexafluorophosphate, hbtu o-benzotriazole-n,n,n',n'-tetramethyl-uronium-hexafluoro-phosphate, n,n,n',n'-tetramethyl-o-1h-benzotriazol-1-yl uronium hexafluorophosphate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-benzotriazole-n,n,n',n'-tetramethyluronium hexafluorophosphate
PubChem CID 2733084
IUPAC Name [benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 200°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C11H16N5O·PF6
Quantity 5 g
Merck Index 15,4654
Solubility Information (10% in DMF) Clear colorless
Molecular Weight (g/mol) 379.25
Formula Weight 379.25
Percent Purity 98%
Chemical Name or Material O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • O-(Benzotriazol-1-yl)-N,N,N`,N`-tetramethyluronium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.