missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl-L-serine, 99%

Product Code. 11480507
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480507 5 g 5g
11470507 1 g 1g
11448710 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480507 Supplier Thermo Scientific Alfa Aesar Supplier No. B22393.06
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4726-96-9 | C10H13NO3 | 195.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4726-96-9
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00065937
InChI Key IDGQXGPQOGUGIX-VIFPVBQESA-N
Synonym o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine
PubChem CID 78457
IUPAC Name (2S)-2-amino-3-phenylmethoxypropanoic acid
SMILES C1=CC=C(C=C1)COCC(C(=O)O)N

Specifications

Melting Point 213°C (decomposition)
Beilstein 2114846
Quantity 5 g
Formula Weight 195.22
Optical Rotation +7° (c=1 in 1N HCl)
Percent Purity 99%
Chemical Name or Material O-Benzyl-L-serine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.