missing translation for 'onlineSavingsMsg'
Learn More

O-Benzylhydroxylamine hydrochloride, 99%

Product Code. 11473637
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473637 5 g 5g
11483637 25 g 25g
11493637 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473637 Supplier Thermo Scientific Alfa Aesar Supplier No. A17393.06
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2687-43-6 | C7H10ClNO | 159.613 g/mol

For use in the formation of nitrones from carbonyl compounds and their use in cycloaddition reactions. Used as a reagent used in the synthesis of hydroxylamines and hydroxyamates. Effective reagent used to prepare α-hydroxybenzylamines from α-hydroxyketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For use in the formation of nitrones from carbonyl compounds and their use in cycloaddition reactions. Used as a reagent used in the synthesis of hydroxylamines and hydroxyamates. Effective reagent used to prepare α-hydroxybenzylamines from α-hydroxyketones.

Solubility
Soluble in DMSO and methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2687-43-6
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.613
MDL Number MFCD00012952
InChI Key HYDZPXNVHXJHBG-UHFFFAOYSA-N
Synonym o-benzylhydroxylamine hydrochloride, benzyloxyamine hydrochloride, o-benzylhydroxylamine hcl, o-benzyl-hydroxylamine hydrochloride, hydroxylamine, o-phenylmethyl-, hydrochloride, hydroxylamine, o-benzyl-, hydrochloride, o-benzyl-hydroxylamine hcl, o-benzyl hydroxylamine hydrochloride, aminooxy methyl benzene hydrochloride, oxybenzylamine, chloride
PubChem CID 102312
IUPAC Name O-benzylhydroxylamine;hydrochloride
SMILES C1=CC=C(C=C1)CON.Cl

Specifications

Melting Point ∼238°C (sublimation)
Quantity 5 g
Beilstein 3595440
Sensitivity Hygroscopic
Solubility Information Soluble in DMSO and methanol.
Formula Weight 159.62
Percent Purity 99%
Chemical Name or Material O-Benzylhydroxylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.