missing translation for 'onlineSavingsMsg'
Learn More

o-Chloromethyltoluene, TRC

Product Code. 30363764
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30363764

Brand: Toronto Research Chemicals TRCC36979050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material o-Chloromethyltoluene
CAS 552-45-4
Molecular Formula C8 H9 Cl
Quantity 50 g
Synonym 1-(Chloromethyl)-2-methylbenzene, 1-Methyl-2-chloromethylbenzene, 2-(Chloromethyl)toluene, 2-Methylbenzyl Chloride, o-Methylbenzyl Chloride, o-Xylyl Chloride, o-Xylyl-α-chloride, α-Chloro-o-xylene, ω-Chloro-o-xylene
InChI Formula InChI=1S/C8H9Cl/c1-7-4-2-3-5-8(7)6-9/h2-5H,6H2,1H3
SMILES Cc1ccccc1CCl
IUPAC Name 1-(chloromethyl)-2-methylbenzene
Molecular Weight (g/mol) 140.61
Formula Weight 140.0393
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.