missing translation for 'onlineSavingsMsg'
Learn More

o-Cresolphthalein complexone tetrasodium salt

Product Code. 11429076
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429076 5 g 5g
11419076 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429076 Supplier Thermo Scientific Alfa Aesar Supplier No. A17941.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 62698-54-8 | C32H28N2Na4O12 | 724.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62698-54-8
Molecular Formula C32H28N2Na4O12
Molecular Weight (g/mol) 724.54
MDL Number MFCD00066371
InChI Key KSMPPVYJZJBIES-UHFFFAOYSA-J
Synonym phthalein complexon sodium salt, o-cresolphthalein complexone tetrasodium salt, sodium 2,2',2,2'-3-oxo-1,3-dihydroisobenzofuran-1,1-diyl bis 2-hydroxy-3-methyl-5,1-phenylene bis methylene bis azanetriyl tetraacetate, phthaleincomplexonsodiumsalt, phthalein complexone sodium salt, glycine, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-5-methyl-3,1-phenylene methylene bis n-carboxymethyl-, sodium salt 1:4, glycine, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-5-methyl-3,1-phenylene methylene bis n-carboxymethyl-, tetrasodium salt, tetrasodium 2,2',2,2'-3-oxo-1,3-dihydro-2-benzofuran-1,1-diyl bis 6-hydroxy-5-methylbenzene-3,1-diyl methanediylnitrilo tetraacetate, tetrasodium 2-5-1-3-bis carboxylatomethyl amino methyl-4-hydroxy-5-methylphenyl-3-oxo-2-benzofuran-1-yl-2-hydroxy-3-methylphenyl methyl carboxylatomethyl amino acetate, tetrasodium 2-5-1-3-bis 2-oxido-2-oxoethyl amino methyl-4-hydroxy-5-methylphenyl-3-oxo-2-benzofuran-1-yl-2-hydroxy-3-methylphenyl methyl-2-oxido-2-oxoethyl amino acetate
PubChem CID 112916
IUPAC Name tetrasodium;2-[[5-[1-[3-[[bis(carboxylatomethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxylatomethyl)amino]acetate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1

Specifications

Quantity 5 g
Formula Weight 724.54
Chemical Name or Material o-Cresolphthalein complexone tetrasodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.