missing translation for 'onlineSavingsMsg'
Learn More

O-Cresolsulfonphthaleindi-(Methyl-Iminodiacetic Acid) Sodium Salt, Pure, special reagent for metals, Fisher Chemical™

Product Code. 10010060 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10010060

Brand: Fisher Chemical C/7480/44

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1611-35-4
Molecular Formula C31H32N2O13S
Molecular Weight (g/mol) 672.66
MDL Number 11493
InChI Key ORZHVTYKPFFVMG-UHFFFAOYSA-N
IUPAC Name 2-[({5-[3-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
SMILES CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1

Specifications

Color Purple
Physical Form Solid
Quantity 5 g
Packaging Glass Jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.