missing translation for 'onlineSavingsMsg'
Learn More

O-Desethyl Candesartan Cilexetil, TRC

Product Code. 17939744
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17939744 10 mg 10mg
17987904 1 mg 1mg
17910085 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17939744 Supplier Toronto Research Chemicals Supplier No. TRCD28885510MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869631-11-8
Molecular Formula C31 H30 N6 O6
Molecular Weight (g/mol) 582.61
Synonym O-Desethyl Candesartan Cilexetil, (1RS)-1-[[(Cyclohexyloxy)carbonyl]-oxy]ethyl 2-Oxo-3-[[2'-(1H-tetrazol-5-yl-)-biphenyl-4-yl]methyl]-2,3-dihydro-1H-benzimidazole-4-carboxylate, 1H-Benzimidazole-4-carboxylic acid, 2,3-dihydro-2-oxo-3-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, 1H-Benzimidazole-4-carboxylic acid, 2,3-dihydro-2-oxo-3-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester (9CI), Candesartan Cilexetil Imp. B (EP)
IUPAC Name 1-cyclohexyloxycarbonyloxyethyl 2-oxo-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]-1H-benzimidazole-4-carboxylate
SMILES CC(OC(=O)OC1CCCCC1)OC(=O)c2cccc3NC(=O)N(Cc4ccc(cc4)c5ccccc5c6nnn[nH]6)c23

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C31H30N6O6/c1-19(42-31(40)43-22-8-3-2-4-9-22)41-29(38)25-12-7-13-26-27(25)37(30(39)32-26)18-20-14-16-21(17-15-20)23-10-5-6-11-24(23)28-33-35-36-34-28/h5-7,10-17,19,22H,2-4,8-9,18H2,1H3,(H,32,39)(H,33,34,35,36)
Formula Weight 582.2227
Percent Purity >95%
Chemical Name or Material O-Desethyl Candesartan Cilexetil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.