missing translation for 'onlineSavingsMsg'
Learn More

O-Desmethyl Brinzolamide, TRC

Product Code. 30133840
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133840 50 mg 50mg
30004513 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133840 Supplier Toronto Research Chemicals Supplier No. TRCB67761050MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186377-56-0
Molecular Formula C11 H19 N3 O5 S3
Molecular Weight (g/mol) 369.4807
Synonym 2H-Thieno[3,2-e]-1,2-thiazine-6-sulfonamide, 4-(ethylamino)-3,4-dihydro-2-(3-hydroxypropyl)-, 1,1-dioxide, (4R)-, 2H-Thieno[3,2-e]-1,2-thiazine-6-sulfonamide, 4-(ethylamino)-3,4-dihydro-2-(3-hydroxypropyl)-, 1,1-dioxide, (R)- (9CI), (4R)-4-(Ethylamino)-3,4-dihydro-2-(3-hydroxypropyl)-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-Dioxide, (R)-4-(Ethylamino)-3,4-dihydro-2-(3-hydroxypropyl)-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-Dioxide, Brinzolamide O-Desmethyl Impurity
IUPAC Name (4R)-4-(ethylamino)-2-(3-hydroxypropyl)-1,1-dioxo-3,4-dihydrothieno[3,2-e]thiazine-6-sulfonamide
SMILES CCN[C@H]1CN(CCCO)S(=O)(=O)c2sc(cc12)S(=O)(=O)N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H19N3O5S3/c1-2-13-9-7-14(4-3-5-15)22(18,19)11-8(9)6-10(20-11)21(12,16)17/h6,9,13,15H,2-5,7H2,1H3,(H2,12,16,17)/t9-/m0/s1
Formula Weight 369.0487
Chemical Name or Material O-Desmethyl Brinzolamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.