missing translation for 'onlineSavingsMsg'
Learn More

O-Desmethyl N-Methyl Lacosamide, TRC

Product Code. 30368580
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368580 100 mg 100mg
30355414 50 mg 50mg
30360658 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368580 Supplier Toronto Research Chemicals Supplier No. TRCD294770100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C13H18N2O3
Molecular Weight (g/mol) 250.29
Synonym (R)-2-Acetamido-N-benzyl-3-hydroxy-N-methylpropanamide
IUPAC Name (R)-2-acetamido-N-benzyl-3-hydroxy-N-methylpropanamide
SMILES O=C([C@H](NC(C)=O)CO)N(C)CC1=CC=CC=C1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H18N2O3/c1-10(17)14-12(9-16)13(18)15(2)8-11-6-4-3-5-7-11/h3-7,12,16H,8-9H2,1-2H3,(H,14,17)/t12-/m1/s1
Formula Weight 250.13
Chemical Name or Material O-Desmethyl N-Methyl Lacosamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.