missing translation for 'onlineSavingsMsg'
Learn More

O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate, 97%

Product Code. 11395015
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11395015 5 g 5g
11385015 1 g 1g
11305025 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11395015 Supplier Thermo Scientific Alfa Aesar Supplier No. L18496.06
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 136849-72-4 | C10H17BF4N4O3 | 328.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136849-72-4
Molecular Formula C10H17BF4N4O3
Molecular Weight (g/mol) 328.07
MDL Number MFCD00192127 MFCD00192127
InChI Key FPQVGDGSRVMNMR-UHFFFAOYSA-N
Synonym totu, o-cyano ethoxycarbonyl methylenamino-1,1,3,3-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethylenamino n,n,n',n'-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethylenamino-n,n,n',n'-tetra methyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethyleneamino-n,n,n',n'-tetramethyluronium bf4, e-2-1-cyano-2-ethoxy-2-oxoethylideneamino-1,1,3,3-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethylenamino-n,n,n'n'-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethyleneamino-n,n,n',n'-tetramethyl uronium tetrafluoroborate, o-ethoxycarboxyl cyanomethyleneamino-n,n,n',n'-tetramethyluronium tetrafluoroborate
PubChem CID 9601231
IUPAC Name [[(E)-(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-(dimethylamino)methylidene]-dimethylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCOC(=O)C(=NOC(N(C)C)=[N+](C)C)C#N

Specifications

Melting Point 143°C to 146°C (decomposition)
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 328.08
Percent Purity 97%
Chemical Name or Material O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.