missing translation for 'onlineSavingsMsg'
Learn More

O-Formyl Acetate (Technical Grade), TRC

Product Code. 30408268
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408268 5 g -
30393178 500 mg 500mg
30390323 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408268 Supplier Toronto Research Chemicals Supplier No. TRCF6934505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2258-42-6
Molecular Formula C3H4O3
Molecular Weight (g/mol) 88.06
Synonym Acetic-formic Anhydride, Acetic-formic mixed Anhydride, Acetoformic Anhydride, Acetyl Formate, Acetyl Formyl Anhydride, Formic-acetic Anhydride, Formyl Acetic Anhydride, Formylacetyl Anhydride
IUPAC Name formyl acetate
SMILES CC(=O)OC=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H4O3/c1-3(5)6-2-4/h2H,1H3
Formula Weight 88.016
Chemical Name or Material O-Formyl Acetate (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.