missing translation for 'onlineSavingsMsg'
Learn More

O-Methyl-L-lactic Acid Ethyl Ester, TRC

Product Code. 30466169
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30466169 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 30466169

Brand: Toronto Research Chemicals TRCM315250500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-Methyl-L-lactic Acid Ethyl Ester
CAS 41918-08-5
Molecular Formula C6 H12 O3
Quantity 500 mg
Synonym 2-Methoxypropionic Acid Ethyl Ester, Ethyl 2-Methoxypropanoate, Ethyl (2S)-2-methoxypropanoate
InChI Formula InChI=1S/C6H12O3/c1-4-9-6(7)5(2)8-3/h5H,4H2,1-3H3/t5-/m0/s1
SMILES CCOC(=O)[C@H](C)OC
IUPAC Name ethyl (2S)-2-methoxypropanoate
Molecular Weight (g/mol) 132.16
Formula Weight 132.0786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.