missing translation for 'onlineSavingsMsg'
Learn More

O-Methyl Mycophenolate Mofetil (EP Impurity D), TRC

Product Code. 30477281
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477281 1 mg 1mg
30476209 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477281 Supplier Toronto Research Chemicals Supplier No. TRCM3205451MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1322681-37-7
Molecular Formula C24 H33 N O7
Molecular Weight (g/mol) 447.52
Synonym 4-Hexenoic acid, 6-(1,3-dihydro-4,6-dimethoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-, 2-(4-morpholinyl)ethyl ester, (4E)-, 2-(Morpholin-4-yl)ethyl (4E)-6-(4,6-Dimethoxy-7-methyl-3-oxo-1,3-dihydro-isobenzofuran-5-yl)-4-methylhex-4-enoate, Mycophenolate Mofetil Imp. D (EP)
IUPAC Name 2-morpholin-4-ylethyl (E)-6-(4,6-dimethoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
SMILES COc1c(C)c2COC(=O)c2c(OC)c1C\C=C(/C)\CCC(=O)OCCN3CCOCC3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C24H33NO7/c1-16(6-8-20(26)31-14-11-25-9-12-30-13-10-25)5-7-18-22(28-3)17(2)19-15-32-24(27)21(19)23(18)29-4/h5H,6-15H2,1-4H3/b16-5+
Formula Weight 447.2257
Chemical Name or Material O-Methyl Mycophenolate Mofetil (EP Impurity D)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.