missing translation for 'onlineSavingsMsg'
Learn More

O-Methylisourea hemisulfate, 94%

Product Code. 10591481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10591481 25 g 25g
10347990 250 g 250g
10423645 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10591481 Supplier Thermo Scientific Acros Supplier No. 233030250
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 52328-05-9 | 0·5 H2SO4 | 123.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52328-05-9
Molecular Formula 0·5 H2SO4
Molecular Weight (g/mol) 123.12
MDL Number MFCD00040594
InChI Key QSCPQKVWSNUJLJ-UHFFFAOYSA-N
Synonym o-methylisourea hemisulfate, 2-methylisouronium sulfate, o-methylisourea hemisulfate salt, bis methoxymethanimidamide ; sulfuric acid, carbamimidic acid, methyl ester, sulfate 2:1, omi™, o-methylpseudourea sulfate, 0-methylisourea hemisulfate, o-methylisourea hemisulphate, bis 2-methylisouronium sulphate
PubChem CID 11357018
IUPAC Name methyl carbamimidate;sulfuric acid
SMILES COC(=N)N.COC(=N)N.OS(=O)(=O)O

Specifications

CAS Min % 92.0
CAS Max % 100.0
Melting Point 147°C to 153°C
Assay Percent Range 94%
Packaging Plastic bottle
Linear Formula 0·5H2SO4
Quantity 25 g
Solubility Information Solubility in water: 1000g/L water (20°C)
Formula Weight 123.12
Percent Purity 94%
Chemical Name or Material O-Methylisourea hemisulfate, 94%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.