missing translation for 'onlineSavingsMsg'
Learn More

O-Methylisourea hemisulfate, 99%

Product Code. 11429527
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429527 250 g 250g
11419527 50 g 50g
11409527 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11429527 Supplier Thermo Scientific Alfa Aesar Supplier No. B22036.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 52328-05-9 | C4H14N4O6S | 246.238 g/mol

O-Methylisoureahemisulfate is used for the synthesis of fluorouracil antineoplastic and anthelmintic imidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Methylisoureahemisulfate is used for the synthesis of fluorouracil antineoplastic and anthelmintic imidazole.

Solubility
Soluble in organic solvents, water, methanol and ethanol

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52328-05-9
Molecular Formula C4H14N4O6S
Molecular Weight (g/mol) 246.238
MDL Number MFCD00040594
InChI Key QSCPQKVWSNUJLJ-UHFFFAOYSA-N
Synonym o-methylisourea hemisulfate, 2-methylisouronium sulfate, o-methylisourea hemisulfate salt, bis methoxymethanimidamide ; sulfuric acid, carbamimidic acid, methyl ester, sulfate 2:1, omi™, o-methylpseudourea sulfate, 0-methylisourea hemisulfate, o-methylisourea hemisulphate, bis 2-methylisouronium sulphate
PubChem CID 11357018
IUPAC Name methyl carbamimidate;sulfuric acid
SMILES COC(=N)N.COC(=N)N.OS(=O)(=O)O

Specifications

Melting Point 163°C to 167°C
Quantity 250 g
Sensitivity Hygroscopic
Solubility Information Soluble in organic solvents,water,methanol and ethanol
Formula Weight 123.12
Percent Purity 99%
Chemical Name or Material O-Methylisourea hemisulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.