missing translation for 'onlineSavingsMsg'
Learn More

O-Methylisourea Hemisulfate, TRC

Product Code. 30473674
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473674 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473674 Supplier Toronto Research Chemicals Supplier No. TRCM3132905G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 52328-05-9
Molecular Formula C2H6N2O.HO2S0.5
Quantity 5 g
Synonym Methoxyformamidine Sulfate, O-Methylisourea Hydrogen Sulfate, O-Methylisouronium Sulfate, O-Methylpseudourea Sulfate, Carbamimidic Acid Methyl Ester Sulfate
InChI Formula InChI=1S/2C2H6N2O.H2O4S/c2*1-5-2(3)4;1-5(2,3)4/h2*1H3,(H3,3,4);(H2,1,2,3,4)
SMILES COC(=N)N.COC(=N)N.OS(=O)(=O)O
IUPAC Name methyl carbamimidate;sulfuric acid
Molecular Weight (g/mol) 246.25
Formula Weight 246.0634
Chemical Name or Material O-Methylisourea Hemisulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.