missing translation for 'onlineSavingsMsg'
Learn More

o-Nitrofluorobenzene, TRC

Product Code. 30138473
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138473 100 g 100g
30132696 50 g 50g
30003672 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138473 Supplier Toronto Research Chemicals Supplier No. TRCN495920100G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1493-27-2
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.1
Synonym 1-Fluoro-2-nitrobenzene, 2-Fluoro-1-nitrobenzene, 2-Fluoronitrobenzene, 2-Nitro-1-fluorobenzene, 2-Nitrofluorobenzene, 4-Fluoro-3-nitrobenzene, NSC 51869, o-Fluoronitrobenzene;
IUPAC Name 1-fluoro-2-nitrobenzene
SMILES [O-][N+](=O)c1ccccc1F

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H4FNO2/c7-5-3-1-2-4-6(5)8(9)10/h1-4H
Formula Weight 141.0226
Chemical Name or Material o-Nitrofluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.