missing translation for 'onlineSavingsMsg'
Learn More

O,O-Dimethyl Dithiophosphate (~90%), TRC

Product Code. 30007753
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007753 500 mg 500mg
30006656 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007753 Supplier Toronto Research Chemicals Supplier No. TRCD465742500MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 756-80-9
Molecular Formula C2H7O2PS2
Molecular Weight (g/mol) 158.18
Synonym Methyl Phosphorodithioate, O,O-Dimethyl Dithiophosphoric Acid, O,O-Dimethyl Phosphorodithioate, O,O-Dimethyl Phosphorodithioic Acid, O,O'-Dimethyl Hydrogen Dithiophosphate, O,O-Dimethyl S-Hydrogen Phosphorodithioate
IUPAC Name O,O-dimethyl S-hydrogen phosphorodithioate
SMILES S=P(OC)(S)OC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C2H7O2PS2/c1-3-5(6,7)4-2/h1-2H3,(H,6,7)
Formula Weight 157.96
Chemical Name or Material O,O-Dimethylphosphorodithioic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.