missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O,O-Dimethyl Methylphosphonate, TRC
Specifications
Specifications
| Chemical Name or Material | Dimethyl methylphosphonate |
| CAS | 756-79-6 |
| Molecular Formula | C3 H9 O3 P |
| Quantity | 50 g |
| Synonym | Reoflam DMMP, DMMP, Dimethoxymethyl Phosphine Oxide, Dimethyl Methanephosphonate, Dimethyl Methylphosphonate, Fran TF 2000, Fyrol DMMP, Metaran, Methanephosphonic Acid Dimethyl Ester, Methylphosphonic Acid Dimethyl Ester, NSC 62240, Methylphosphonic Acid Dimethyl Ester, p-Methylphosphonic Acid Dimethyl Ester |
| InChI Formula | InChI=1S/C3H9O3P/c1-5-7(3,4)6-2/h1-3H3 |
| SMILES | COP(=O)(C)OC |
| IUPAC Name | [methoxy(methyl)phosphoryl]oxymethane |
| Molecular Weight (g/mol) | 124.076 |
| Formula Weight | 124.0289 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?