missing translation for 'onlineSavingsMsg'
Learn More

(-)-O,O'-Dipivaloyl-L-tartaric acid, 98+%, Thermo Scientific™

Product Code. 15482458
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482458 5 g 5g
15472458 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15482458 Supplier Thermo Scientific Alfa Aesar Supplier No. H56032.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65259-81-6
Molecular Formula C14H22O8
Molecular Weight (g/mol) 318.32
MDL Number MFCD00066535
InChI Key UFHJEZDFEHUYCR-YZYOREDDNA-N
PubChem CID 16212279
IUPAC Name (2R,3R)-2,3-bis[(2,2-dimethylpropanoyl)oxy]butanedioic acid
SMILES CC(C)(C)C(=O)O[C@H]([C@@H](OC(=O)C(C)(C)C)C(O)=O)C(O)=O

Specifications

Melting Point 127°C to 132°C
Beilstein 2008171
Synonym +-dipivaloyl-d-tartaric acid, 2s,3s-2,3-bis pivaloyloxy succinic acid, d-+-dipivaloyl tartaric acid, pubchem8173, --bis 2,2-dimethylpropionyl-l-tartaric acid, dipivaloyl-l-tartaric acid, +-dipivaloyl-d-tartaricacid, +-dpta, ?-o,o'-di-pivaloyl-l-tartaric acid, +-o,o'-di-pivaloyl-d-tartaric acid
Formula Weight 318.32
Percent Purity ≥98%
Quantity 5 g
Optical Rotation −24° (c=1.7 in Dioxane)
Sensitivity Hygroscopic
Chemical Name or Material (-)-O,O'-Dipivaloyl-L-tartaric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.