missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O,O'-Dipropanal Ganciclovir, TRC
Specifications
Specifications
| Chemical Name or Material | 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]propane-1,3-diyl Dipropanoate (Ganciclovir Dipropionate) |
| CAS | 86357-20-2 |
| Molecular Formula | C15 H21 N5 O6 |
| Quantity | 250 mg |
| Synonym | 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]propane-1,3-diyl Dipropanoate, Ganciclovir Imp. I (EP), Ganciclovir Dipropionate |
| InChI Formula | InChI=1S/C15H21N5O6/c1-3-10(21)24-5-9(6-25-11(22)4-2)26-8-20-7-17-12-13(20)18-15(16)19-14(12)23/h7,9H,3-6,8H2,1-2H3,(H3,16,18,19,23) |
| SMILES | CCC(=O)OCC(COC(=O)CC)OCn1cnc2C(=O)NC(=Nc12)N |
| IUPAC Name | [2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]-3-propanoyloxypropyl] propanoate |
| Molecular Weight (g/mol) | 367.36 |
| Formula Weight | 367.1492 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?