missing translation for 'onlineSavingsMsg'
Learn More

O,O,S-Trimethyl Dithiophosphate, TRC

Product Code. 18062920
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18062920 50 mg 50mg
18031910 100 mg 100mg
18030420 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18062920 Supplier Toronto Research Chemicals Supplier No. TRCT79669050MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2953-29-9
Molecular Formula C3 H9 O2 P S2
Molecular Weight (g/mol) 172.21
Synonym Phosphorodithioic acid, O,O,S-trimethyl ester, Methyl phosphorodithioate ((MeO)2(MeS)PS) (6CI), Methyl phosphorodithioate (7CI), O,O,S-Trimethyl dithiophosphate, O,O,S-Trimethyl phosphorodithioate
IUPAC Name dimethoxy-methylsulfanyl-sulfanylidene-λ^{5}-phosphane
SMILES COP(=S)(OC)SC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C3H9O2PS2/c1-4-6(7,5-2)8-3/h1-3H3
Formula Weight 171.9782
Percent Purity >95%
Chemical Name or Material O,O,S-Trimethyldithiophosphate
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.