missing translation for 'onlineSavingsMsg'
Learn More

O-Pentylresorufin, TRC

Product Code. 30427327
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427327 1 mg -
30422953 10 mg 10mg
30416533 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427327 Supplier Toronto Research Chemicals Supplier No. TRCP2842001MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87687-03-4
Molecular Formula C17 H17 N O3
Molecular Weight (g/mol) 283.32
Synonym 7-(Pentyloxy)-3H-phenoxazin-3-one, 7-Pentoxyphenoxazone, 7-Pentoxyresorufin
IUPAC Name 7-pentoxyphenoxazin-3-one
SMILES CCCCCOc1ccc2N=C3C=CC(=O)C=C3Oc2c1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H17NO3/c1-2-3-4-9-20-13-6-8-15-17(11-13)21-16-10-12(19)5-7-14(16)18-15/h5-8,10-11H,2-4,9H2,1H3
Formula Weight 283.1208
Chemical Name or Material O-Pentylresorufin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.