missing translation for 'onlineSavingsMsg'
Learn More

o-Phenylenediamine, Fisher BioReagents

Product Code. 10718705
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Amber Glass
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10718705 250 g Amber Glass 250g
10422305 50 g Amber Glass 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10718705 Supplier Fisher BioReagents Supplier No. BP2537250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Free Base

  • Commonly used as a substrate for horseradish peroxidase (HRP) in ELISAs
  • When oxidized, OPD produces a color change that is detectable by spectrophotometry
  • Color intensity will increase over time as OPD is oxidzed by peroxidase
  • A chromogenic peroxidase substrate useful for ELISA and other enzyme immunoassays
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-54-5
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
InChI Key GEYOCULIXLDCMW-UHFFFAOYSA-N
Synonym o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine
PubChem CID 7243
ChEBI CHEBI:34043
IUPAC Name benzene-1,2-diamine
SMILES C1=CC=C(C(=C1)N)N

Specifications

Melting Point 100°C
Color Cream
Boiling Point 256°C
Packaging Amber Glass
Linear Formula C6H4(NH2)2
Quantity 250 g
Merck Index 15, 7396
Formula Weight 108.1
Physical Form Solid
Chemical Name or Material o-Phenylenediamine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • o-Phenylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.