missing translation for 'onlineSavingsMsg'
Learn More

O-Phosphorylethanolamine, 98%

Product Code. 10039171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10039171 5 g Glass Bottle 5g
10742063 25 g Glass Bottle 25g
15239876 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10039171 Supplier Thermo Scientific Acros Supplier No. 367420050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1071-23-4 | C2H8NO4P | 141.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-23-4
Molecular Formula C2H8NO4P
Molecular Weight (g/mol) 141.06
MDL Number MFCD00008178
InChI Key SUHOOTKUPISOBE-UHFFFAOYSA-N
Synonym o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate
PubChem CID 1015
ChEBI CHEBI:17553
IUPAC Name 2-aminoethyl dihydrogen phosphate
SMILES C(COP(=O)(O)O)N

Specifications

Melting Point 241.0°C to 243.0°C
Assay Percent Range 97.5 % min. (Titration with HClO4)
Packaging Glass Bottle
Linear Formula H2NCH2CH2OP(O)(OH)2
Quantity 5 g
Beilstein 04,IV,1415
Solubility Information Solubility in water: soluble.
Formula Weight 141.06
Percent Purity 98%
Chemical Name or Material O-Phosphorylethanolamine, 98%
compliance-icons
Product Identifier
  • O-Phosphorylethanolamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.