missing translation for 'onlineSavingsMsg'
Learn More

o-Phthaloyl dichloride, 90%, remainder mainly phthalic acid, Thermo Scientific Chemicals

Product Code. 10745611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10745611 5 mL Glass bottle 5mL
10031400 100 mL Glass bottle 100mL
10157753 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10745611 Supplier Thermo Scientific Acros Supplier No. 131110050
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 88-95-9 | Molecular Formula: C8H4Cl2O2 | Molecular Weight: 203.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-95-9
MDL Number MFCD00000666
InChI Key FYXKZNLBZKRYSS-UHFFFAOYSA-N
Synonym phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride
PubChem CID 6955
IUPAC Name benzene-1,2-dicarbonyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl

Specifications

Melting Point 12.0°C
Color Yellow
Density 1.4000g/mL
Boiling Point 269.0°C to 271.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Molecular Formula C8H4Cl2O2
Linear Formula C6H4(COCl)2
Refractive Index 1.5674 to 1.5694
Quantity 5 mL
Beilstein 09, 805
Fieser 01,882; 14,263
Merck Index 15, 7487
Specific Gravity 1.4
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether, decomposes in alcohol
Molecular Weight (g/mol) 203.02
Formula Weight 203.02
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material o-Phthaloyl dichloride, Remainder mainly phthalic acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • o-Phthaloyl dichloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1,2-Benzenedicarbonyl dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.