missing translation for 'onlineSavingsMsg'
Learn More

o-Phthaloyl Dichloride, TRC

Product Code. 30419695
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419695 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419695 Supplier Toronto Research Chemicals Supplier No. TRCP39000025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material o-Phthaloyl Dichloride
CAS 88-95-9
Molecular Formula C8H4Cl2O2
Quantity 25 g
Synonym 1,2-Benzenedicarbonyl Dichloride, Phthaloyl Chloride, 1,2-Bis(chlorocarbonyl)benzene, 1,2-Phthaloyl Dichloride, NSC 44611, Phthalic Acid Dichloride, Phthalic Chloride, Phthalic Dichloride, Phthaloyl Dichloride, Phthalyl Chloride, Phthalyl Dichloride, o-Phthalic Acid Dichloride
InChI Formula InChI=1S/C8H4Cl2O2/c9-7(11)5-3-1-2-4-6(5)8(10)12/h1-4H
SMILES ClC(=O)c1ccccc1C(=O)Cl
IUPAC Name benzene-1,2-dicarbonyl chloride
Molecular Weight (g/mol) 203.02
Formula Weight 201.9588
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.