missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ O-Propargyl-puromycin
GREENER_CHOICE

Product Code. 18775956
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18775956 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18775956 Supplier Tocris Bioscience™ Supplier No. 7391/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puromycin analog for labeling newly synthesized proteins; is incorporated into nascent polypeptides

O-Propargyl-puromycin (OP-puro) is a puromycin analog that is incorporated into nascent polypeptides. It is used for the labeling of newly synthesized proteins in cells and whole animals in vivo. O-Propargyl-puromycin can be click-conjugated to fluorescent azides for visualization by fluorescence microscopy. The product does not require methionine-free conditions. It can also be used in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3 '-[[(2 S)-2-Amino-1-oxo-3-[4-(2-propyn-1-yloxy)phenyl]propyl]amino]-3 '-deoxy-N,N-dimethyladenosine
CAS 1416561-90-4
Quantity 1 mg
Target Other Fluorescent Probes
Molecular Formula C24H29N7O5
Purity 0.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.