missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ohaus™ O-Ring, 250 mL Centrifuge Rack Adaptor Cap, Frontier™ R-S4x250/4MI Swing Out Rotors
Used with centrifuge rack screw cap to contain bucket contents.
Brand: Ohaus™ 30314902
Additional Details : Weight : 1.00000kg
Description
- Suitable for use with 250 mL rack adaptors for R-S4x250/4MI
- Use in conjunction with 250 mL screw cap
Specifications
250 mL Centrifuge Rack Adaptor, Frontier™ R-S4x250/4MI Swing Out Rotors, Screw Cap | |
O-ring, Rack of 250ml Rotor, 10/pk |