Learn More
Bohlender™ O-Ring
Product Code. 10706002
Change view
Click to view available options
Material:
Polytetrafluoroethylene
Unit Size:
Pack of 10
Product Code. Material unitSize
10706002 Polytetrafluoroethylene Pack of 10
1 options
Product Code. 10706002 Supplier Bohlender™ Supplier No. H97510
Only null left
Add to basket
Edge
Add to basket
Specifications
For Use With (Application) 18 Thread GL and 9 x11mm dia. Tube . As gasket for BOLA screw caps with aperture (Cat. No. H 983/ H 984/ H 995 on page 88). Also suitable for GL caps of company Duran Group (formerly schott AG).
Quantity 10/Pk.
Product Type O-Ring
Material Polytetrafluoroethylene
Diameter (Metric) Outside 16 mm
Size 16 O.D x 10 I.D.mm