Learn More
Cytiva O-ring
Product Code. 11785185 Shop All Cytiva Products
Change view
Click to view available options
Size:
11.3 x 2.4mm
27.1 x 1.60 mm
37.47 x 5.34mm
6.75 x 1.78mm
Unit Size:
Each
Pack of 10
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11785185 11.3 x 2.4mm Each
10201640 37.47 x 5.34mm Pack of 5
11570854 27.1 x 1.60 mm Pack of 5
11530854 6.75 x 1.78mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11785185 Supplier Cytiva Supplier No. 18110497
In Stock
Add to basket
Add to basket
Use in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules. Cytiva Life Sciences™ O-rings are made of nitrile rubber.
  • Can be used in aqueous solutions
  • Nearly all organic solvents
  • The following few exceptions: chlorinated hydrocarbons, acetone and otherketones, aliphatic esters and phenol. Solutions containingmore than 10% NaOH, 10% HCl, other strong mineral acidsor 5% acetic acid should not be used
  • Made of Nitrile Rubber (NBR)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2/Pk.
Product Type O-Ring
Material Perfluor-Rubber
Size 11.3 x 2.4mm