Learn More
DWK Life Sciences O-Ring, red, FEP coated, not suitable for desiccators
Product Code. 10098240 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Outside:
75 mm
110 mm
133 mm
157 mm
215 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside unitSize
10098240 110 mm Each
10180151 75 mm Each
10029970 133 mm Each
12646203 157 mm Each
10613602 215 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10098240 Supplier DWK Life Sciences Supplier No. 292224608
Add to basket
Add to basket
Seal is suitable for the tight sealing of flat flange vessels with annular groove.
Flat flange vessels are large-volume reaction vessels consisting of a lower part and a lid. These two parts are held together by a clamping ring. The red O-ring made of silicone is seamlessly coated with FEP and serves as an optimal seal for flat flange vessels with an annular groove. It prevents jamming of the ground surfaces between the base and the lid and is convincing due to its good elasticity in combination with outstanding chemical resistance.
  • Accessories for flat flange vessels, comprising an elastic, silicone core with seamless FEP coating that encloses the ring
  • The combination of these high-quality materials achieves good elasticity in conjunction with outstanding chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Tight closure of flange vessels
Quantity 1 Ea.
Material Silicone, FEP
Diameter (Metric) Outside 110 mm