Learn More
DWK Life Sciences O-Ring, transparent, from silicone (VMQ), not suitable for desiccators
Product Code. 13497068 Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric) Outside:
75 mm
110 mm
133 mm
157 mm
215 mm
Unit Size:
Pack of 5
Product Code. 13497068
Brand: DWK Life Sciences 292255101
Only null left
Add to basket
Add to basket
Seal is suitable for the tight sealing of flat flange vessels with annular groove.
Flat flange vessels are large-volume reaction vessels consisting of a lower part and a lid. These two parts are held together by a clamping ring. The transparent O-ring made of silicone serves as a seal for flat flange vessels with an annular groove. It prevents tilting of the ground surfaces between the lower part and the lid and convinces by its good elasticity. The chemical resistance is limited compared to FEP coated O-rings.
  • Accessories for flat flange vessels from silicone (VMQ), so is highly elastic
  • The chemical resistance of silicone is lower than FEP coated O-rings
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Tight closure of flange vessels
Quantity 5/Pk.
Material Silicone (VMQ)
Diameter (Metric) Outside 133 mm