missing translation for 'onlineSavingsMsg'
Learn More

O,S-Diethyl methylphosphonothioate, 97%, Thermo Scientific Chemicals

Product Code. 11400790
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400790 0.5 g 0.5g
11410790 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400790 Supplier Thermo Scientific Alfa Aesar Supplier No. 044465.02
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

O,S-Diethyl methylphosphonothioate is used as an inhibitor of cholinesterase. It is also used in pharmaceuticals, intermediates and fine chemicals. It also serves as phosphorylating and phosphitylating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2511-10-6
MDL Number MFCD01705977

Specifications

Density 1.094 g/mL
Boiling Point 95°C to 97°C (10 mmHg)
Molecular Formula C5H13O2PS
Refractive Index 1.474
Quantity 0.5 g
UN Number UN3278
Synonym o,s-diethyl methylphosphonothioate, o,s-diethyl methylthiophosphonate, methylphosphonothioic acid o,s-diethyl ester, osdmp, phosphonothioic acid, methyl-, o,s-diethyl ester, phosphonothioic acid, p-methyl-, o,s-diethyl ester, o-ethyl s-ethyl methylphosphonothioate, ethyl ethylsulfanyl methyl phosphinate, acmc-20ak0l, ethoxy ethylthio methyl phosphine oxide
Solubility Information Immiscible with water.
InChI Key AIMOGGQLCCAPQJ-UHFFFAOYNA-N
SMILES CCOP(=O)(C)SCC
IUPAC Name 1-[ethylsulfanyl(methyl)phosphoryl]oxyethane
Molecular Weight (g/mol) 168.19
PubChem CID 520177
Formula Weight 168.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material O,S-Diethyl methylphosphonothioate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.