missing translation for 'onlineSavingsMsg'
Learn More

O-(tert-Butyl)hydroxylamine hydrochloride, 99%

Product Code. 10051644 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051644 1 g Glass bottle 1g
10143144 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10051644 Supplier Thermo Scientific Acros Supplier No. 399680010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39684-28-1 | C4H11NO·HCl | 125.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39684-28-1
Molecular Formula C4H11NO·HCl
Molecular Weight (g/mol) 125.6
InChI Key ZBDXGNXNXXPKJI-UHFFFAOYSA-N
Synonym o-tert-butyl hydroxylamine hydrochloride, o-tert-butylhydroxylamine hydrochloride, o-t-butylhydroxylamine hydrochloride, tert-butoxyamine hydrochloride, o-tert-butyl hydroxylaminehydrochloride, hydroxylamine, o-1,1-dimethylethyl-, hydrochloride, soyheadhmuup@, o-tert-butylhydroxylamine hcl, pubchem19621, t-butoxyamine hydrochloride
PubChem CID 2777906
IUPAC Name O-tert-butylhydroxylamine;hydrochloride
SMILES CC(C)(C)ON.Cl

Specifications

Melting Point 155.0°C
Assay Percent Range 98.5 to 101.5% (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)3CONH2·HCl
Quantity 1 g
Formula Weight 125.6
Percent Purity 99%
Chemical Name or Material O-(tert-Butyl)hydroxylamine hydrochloride

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.