missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-L-serine, 97%

Product Code. 10361380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10361380 1 g 1g
10814341 5 g 5g
10068359 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10361380 Supplier Thermo Scientific Acros Supplier No. 434880010
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 18822-58-7 | C7H15NO3 | 161.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18822-58-7
InChI Key DDCPKNYKNWXULB-YFKPBYRVSA-N
Synonym h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh
PubChem CID 7004940
IUPAC Name (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid
SMILES CC(C)(C)OCC(C(=O)O)N

Specifications

Melting Point 225.2°C to 228.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C7H15NO3
Quantity 1 g
Specific Rotation − 15.50
Molecular Weight (g/mol) 161.2
Formula Weight 161.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material O-tert-Butyl-L-serine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.