missing translation for 'onlineSavingsMsg'
Learn More

o-tert-Butyl-p-cresol, TRC

Product Code. 30340480
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340480 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340480 Supplier Toronto Research Chemicals Supplier No. TRCB69092550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-tert-Butyl-4-methylphenol
CAS 2409-55-4
Molecular Formula C11 H16 O
Quantity 50 g
Synonym 2-(1,1-Dimethylethyl)-4-methylphenol, 2-tert-Butyl-4-cresol, 2-tert-Butyl-4-methylphenol, 4-Methyl-2-(1,1-dimethylethyl)phenol
InChI Formula InChI=1S/C11H16O/c1-8-5-6-10(12)9(7-8)11(2,3)4/h5-7,12H,1-4H3
SMILES Cc1ccc(O)c(c1)C(C)(C)C
IUPAC Name 2-tert-butyl-4-methylphenol
Molecular Weight (g/mol) 164.24
Formula Weight 164.1201
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.