missing translation for 'onlineSavingsMsg'
Learn More

o-Tolyl isocyanate, 99+%

Product Code. 10492825 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10492825 10 g Glass bottle 10g
10697241 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10492825 Supplier Thermo Scientific Acros Supplier No. 292220100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 614-68-6 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-68-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002009
InChI Key VAYMIYBJLRRIFR-UHFFFAOYSA-N
Synonym o-tolyl isocyanate, 2-methylphenyl isocyanate, 2-tolyl isocyanate, o-methylphenyl isocyanate, o-toluene isocyanate, benzene, 1-isocyanato-2-methyl, 2-methylphenylisocyanate, tolyl isocyanate, isocyanic acid, o-tolyl ester, o-tolylisocyanate
PubChem CID 69194
ChEBI CHEBI:60099
IUPAC Name 1-isocyanato-2-methylbenzene
SMILES CC1=CC=CC=C1N=C=O

Specifications

Color Colorless to Yellow
Density 1.0740g/mL
Boiling Point 185°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4NCO
Refractive Index 1.5335 to 1.5355
Quantity 10 g
Beilstein 12, 812
Specific Gravity 1.074
Formula Weight 133.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material o-Tolyl isocyanate, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.