Product Code. 10114953
Shop All Thermo Scientific Chemicals Products
Click to view available options
Packaging:
Glass Bottle
Plastic Drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
unitSize
10114953
10 L
Plastic Drum
10 L
10306600
1 L
Glass Bottle
1 L
10407540
2.5 L
Glass Bottle
2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
10114953
Supplier
Thermo Scientific
Supplier No.
140990100
337.00 € 327.00 € / 10 L Save 10.00 € 2% Off
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
CAS: 95-47-6 | C8H10 | 106.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
95-47-6
Molecular Formula
C8H10
Molecular Weight (g/mol)
106.17
MDL Number
MFCD00008519
InChI Key
CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID
7237
ChEBI
CHEBI:28063
IUPAC Name
1,2-xylene
SMILES
CC1=CC=CC=C1C
Specifications
Melting Point
-25.0°C
Color
Colorless
Density
0.8780g/mL
Quantity
10 L
Flash Point
31°C
Infrared Spectrum
Authentic
Packaging
Plastic Drum
Physical Form
Liquid
Refractive Index
1.5038 to 1.5058
Assay Percent Range
99%
Specific Gravity
0.878
Linear Formula
C6 H4 (CH3 )2
Viscosity
0.81 mPa.s (20°C)
Beilstein
05, 362
Merck Index
15, 10278
Boiling Point
143.0°C to 145.0°C
Solubility Information
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, miscible with ether and most organic solvents
Formula Weight
106.17
Percent Purity
99%
Grade
Pure
Chemical Name or Material
o-Xylene, Pure
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?