missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene, CHROMASOLV™ Plus, for HPLC, 98%, Solstice

Product Code. 15602970
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15602970 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15602970 Supplier Honeywell-Chromasolv Supplier No. 2958841L
  • ONLINE EXCLUSIVE PRICE
    124.00 €
    101.32 € / 1 L
    Save 22.68 €
    18% Off
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CHROMASOLV™ Plus, for HPLC, 98%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -26°C to -23°C (literature)
Absorbance max. 0.01 at 350 nm, max. 0.05 at 325 nm, max. 0.40 at 300 nm, max. 1.00 at 288 nm
CAS 95-47-6
CAS Max % 100.0000%
Density 0.879 g/mL at 20 °C(lit.)
Quantity 1 L
Flash Point 30°C
Packaging Glass Bottle
Molecular Formula C8H10
Linear Formula C6H4(CH3)2
MDL Number MFCD00008519
UN Number UN1307
Beilstein 1815558
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
Boiling Point 143°C to 145°C
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1C
IUPAC Name 1,2-xylene
Molecular Weight (g/mol) 106.17
PubChem CID 7237
ChEBI CHEBI:28063
Formula Weight 106.17
Percent Purity >97.5%
Grade HPLC
Chemical Name or Material o-Xylene, CHROMASOLV™ Plus, for HPLC, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • o-Xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: o-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.