Product Code. 15648200
Click to view available options
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
18 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
15648200
18 L
Stainless Steel Can
18 L
15658200
1 L
Glass Bottle
1 L
15668200
2.5 L
Glass Bottle
2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
15648200
Supplier
Fluka
Supplier No.
X104018L
645.00 € 529.62 € / 18 L Save 115.38 € 17% Off
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Reagent Grade,≥98.0%
≥95% for general laboratory use.
Chemical Identifiers
CAS
95-47-6
Molecular Formula
C8H10
Molecular Weight (g/mol)
106.17
MDL Number
MFCD00008519
InChI Key
CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID
7237
ChEBI
CHEBI:28063
IUPAC Name
1,2-xylene
SMILES
CC1=CC=CC=C1C
Specifications
Density
0.879g/mL at 20°C
Quantity
18 L
Explosion Limit
0.07
Flash Point
31°C (87.8°F)
Packaging
Stainless Steel Can
Linear Formula
C6 H4 (CH3 )2
Vapor Pressure
16 mmHg (37.7°C), 0.1 atm (21.1°C), 7 mmHg (20°C)
UN Number
UN1307
Beilstein
1815558
Formula Weight
106.17g/mol
Percent Purity
≥98%
Grade
Not Available
Chemical Name or Material
o-Xylene
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?