missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene, Spectrophotometric Grade, 96% min

Product Code. 11378407
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378407 4 L 4 L
11368407 1 L 1 L
11358407 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11378407 Supplier Thermo Scientific Alfa Aesar Supplier No. 032471.K7
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 95-47-6 | C8H10 | 106.17 g/mol

o-Xylene is used as a solvent for paints and adhesives, and to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Melting Point -25°C
Color Colorless
Density 0.877 g/mL
Quantity 4 L
Flash Point 32°C (89°F)
Physical Form Liquid
Refractive Index 1.505
Linear Formula (CH3)2C6H4
UN Number UN1307
Beilstein 1815558
Merck Index 14,10081
Boiling Point 143°C to 145°C
Solubility Information Sparingly soluble in water. (0.2g/L)
Formula Weight 106.17
Percent Purity ≤96%
Grade Spectrophotometry
Chemical Name or Material o-Xylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.