Learn More
Motic™ Objective, Achromatic EA 20X/0.40
Product Code. 15455102
Change view
Click to view available options
Magnification Power:
20X
Unit Size:
Each
Product Code. Magnification Power unitSize
15455102 20X Each
1 options
Product Code. 15455102 Supplier Motic™ Supplier No. 12180163
Designed with chromatic aberration correction for 2 colors. Motic™ Objective, Achromatic EA 20X/0.40 is spherically corrected for one color.
  • Achromatic.
  • Chromatic aberration correction for 2 colors.
  • Spherical correction for 1 color.
Specifications
Type Achromat Objective
Description NA: 0.8
Focus Distance 0.80 mm WD
For Use With (Equipment) PM Series, DM1802-A, SERIES 18, Series 28 Microscopes
Magnification Power 20X