Learn More
Zeiss™ Objective Plan
Product Code. 13469166
Change view
Click to view available options
Type:
Objective Plan S
Unit Size:
Each
Product Code. Type Unit Size
13469166 Objective Plan S Each
1 options
Product Code. 13469166 Supplier Zeiss™ Supplier No. 4352000000000
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Benefit from the long experience of Zeiss in building world class optics to find the objectives dedicated for your application
Compatible Microscopes:
  • SteREO Discovery.V12
  • SteREO Discovery.V20
  • SteREO Discovery.V8
  • SteREO Lumar.V12
Specifications
Type Objective Plan S
Focus Distance 81 mm
For Use With (Equipment) Zeiss Microscopes
Magnification Power 1X
Quantity 1 Ea.