missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCIL/CLEC2d Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3376-SP
This item is not returnable.
View return policy
Description
OCIL/CLEC2d Polyclonal specifically detects OCIL/CLEC2d in Mouse samples. It is validated for Western Blot.Specifications
| OCIL/CLEC2d | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLAXLLT-1, CLEC2D, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1, LLT1Osteoclast inhibitory lectin, OCILmember D | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29121 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q91V08 | |
| CLEC2D | |
| E. coli-derived recombinant mouse OCIL/CLEC2d Ala65-Ser207 Accession # Q91V08 | |
| 25 μg | |
| Primary | |
| Detects mouse OCIL/CLEC2d in direct ELISAs and Western blots. In Western blots, approximately 35% cross-reactivity with recombinant mouse OCILRP2 is observed and less than 2% cross-reactivity with recombinant human OCIL is observed. | |
| Mouse | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?