missing translation for 'onlineSavingsMsg'
Learn More

Oclacitinib maleate, MedChemExpress

Product Code. 16488437
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488437 5 mg 5mg
16441917 50 mg 50mg
16416236 100 mg 100mg
16405768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488437 Supplier Medchem Express Supplier No. HY13577A5mg
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oclacitinib maleate (PF-03394197 maleate) is a novel JAK inhibitor. Oclacitinib maleate (PF-03394197 maleate) is most potent at inhibiting JAK1 (IC50=10 nM).

Oclacitinib maleate (PF-03394197 maleate) is a novel JAK inhibitor. Oclacitinib maleate (PF-03394197 maleate) is most potent at inhibiting JAK1 (IC50=10 nM).

Specifications

Chemical Name or Material Oclacitinib maleate
CAS 1640292-55-2
Color White
Purity Grade Notes Research
Molecular Formula C19H27N5O6S
Quantity 5 mg
Synonym PF-03394197 maleate
Solubility Information DMSO : 250 mg/mL (551.26 mM; Need ultrasonic) ∣H2O : 5 mg/mL (11.03 mM; Need ultrasonic)
SMILES O=S(C[C@H]1CC[C@H](N(C)C2=C3C(NC=C3)=NC=N2)CC1)(NC)=O.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 453.51
Formula Weight 453.51
Percent Purity 99.3%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.